Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106209.1 | NP_001099679.1 | 1 - 2 | 309 | 88 | |
XM_006227918.2 | XP_006227980.1 | 3 - 4 | 1209 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |