Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 1 - 2 | 138 | 75 | |
XM_008766750.1 | XP_008764972.1 | 1 - 2 | 183 | 75 | ||
GenBank mRNA | AF482705.1 | - | 1 - 2 | 138 | 75 | |
BC086988.1 | - | 1 - 2 | 149 | 7192831 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |