Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080096.1 | NP_001073565.1 | 11 - 12 | 1806 | 73 | |
XM_006244311.2 | XP_006244373.1 | 11 - 12 | 2315 | 73 | ||
XM_006244312.2 | XP_006244374.1 | 11 - 12 | 2317 | 73 | ||
GenBank mRNA | BC129110.1 | - | 11 - 12 | 1806 | 7111052 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |