Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080096.1 | NP_001073565.1 | 7 - 8 | 989 | 93 | |
XM_006244311.2 | XP_006244373.1 | 7 - 8 | 1498 | 93 | ||
XM_006244312.2 | XP_006244374.1 | 7 - 8 | 1500 | 93 | ||
GenBank mRNA | BC129110.1 | - | 7 - 8 | 989 | 7111052 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |