Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085406.1 | NP_001078875.1 | 3 - 4 | 523 | 85 | |
XM_008766773.1 | XP_008764995.1 | 4 - 5 | 814 | 85 | ||
GenBank mRNA | BC101893.1 | - | 2 - 3 | 351 | 85 | |
BC158552.1 | - | 3 - 4 | 523 | 9098910 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |