Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031239.2 | NP_112518.1 | 6 - 7 | 768 | 82 | |
GenBank mRNA | AB008161.1 | - | 6 - 7 | 768 | 82 | |
AF009602.1 | - | 6 - 7 | 753 | 82 | ||
BC070893.1 | - | 6 - 7 | 764 | 7099223 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |