Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773660.1 | XP_008771882.1 | 1 - 2 | 180 | 150 | |
XM_008773661.1 | XP_008771883.1 | 1 - 2 | 180 | 150 | ||
XM_008773662.1 | XP_008771884.1 | 1 - 2 | 180 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |