Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773660.1 | XP_008771882.1 | 2 - 3 | 364 | 99 | |
XM_008773661.1 | XP_008771883.1 | 2 - 3 | 364 | 99 | ||
GenBank mRNA | FQ227877.1 | - | 1 - 2 | 103 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |