Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007558.1 | NP_001007559.1 | 91 | 109 | ||
XM_008766767.1 | XP_008764989.1 | 85 | 109 | |||
GenBank mRNA | AY686633.1 | - | 91 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |