Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053521.1 | NP_445973.1 | 1 - 2 | 170 | 123 | |
GenBank mRNA | AB030947.1 | - | 1 - 2 | 170 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |