Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147206.2 | NP_671739.2 | 1 - 2 | 163 | 113 | |
GenBank mRNA | BC086985.1 | - | 1 - 2 | 163 | 7125835 | 113 |
U60085.2 | - | 1 - 2 | 139 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |