Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013957.1 | NP_001013979.1 | 4 - 5 | 437 | 68 | |
XM_006256707.1 | XP_006256769.1 | 4 - 5 | 411 | 68 | ||
XM_006256708.2 | XP_006256770.1 | 3 - 4 | 312 | 68 | ||
GenBank mRNA | BC059163.1 | - | 4 - 5 | 437 | 6889015 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |