Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257420.2 | XP_006257482.1 | 10 - 11 | 2910 | 110 | |
XM_006257421.1 | XP_006257483.1 | 11 - 12 | 2597 | 110 | ||
XM_006257422.1 | XP_006257484.1 | 10 - 11 | 2448 | 110 | ||
XM_006257423.2 | XP_006257485.1 | 9 - 10 | 2761 | 110 | ||
XM_006257424.2 | XP_006257486.1 | 8 - 9 | 1827 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |