Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053379.3 | NP_445831.3 | 1 - 2 | 43 | 88 | |
XM_006257383.2 | XP_006257445.1 | 1 - 2 | 261 | 88 | ||
XM_006257384.2 | XP_006257446.1 | 1 - 2 | 260 | 88 | ||
XM_006257385.2 | XP_006257447.1 | 1 - 2 | 261 | 88 | ||
XM_006257386.2 | XP_006257448.1 | 1 - 2 | 261 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |