Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133538.2 | NP_598222.2 | 4 - 5 | 462 | 194 | |
XM_006257427.2 | XP_006257489.1 | 3 - 4 | 330 | 194 | ||
XM_006257428.2 | XP_006257490.1 | 3 - 4 | 448 | 194 | ||
XM_008773388.1 | XP_008771610.1 | 3 - 4 | 602 | 194 | ||
XM_008773389.1 | XP_008771611.1 | 4 - 5 | 489 | 194 | ||
GenBank mRNA | AF448818.1 | - | 4 - 5 | 462 | 194 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |