Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031808.1 | NP_113996.1 | 3 - 4 | 459 | 109 | |
GenBank mRNA | AF067793.1 | - | 3 - 4 | 459 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |