Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ct55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_228664.5 | XP_228664.3 | 1 - 2 | 251 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |