Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159656.1 | NP_001153128.1 | 4 - 5 | 1181 | 66 | |
GenBank mRNA | AF107845.1 | - | 4 - 5 | 1140 | 66 | |
L10326.1 | - | 3 - 4 | 277 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |