Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762615.1 | XP_008760837.1 | 4 - 5 | 509 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |