Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198755.1 | NP_942050.1 | 561 | 83 | ||
XM_006235502.2 | XP_006235564.1 | 5 - 6 | 690 | 83 | ||
XM_006235503.2 | XP_006235565.1 | 4 - 5 | 609 | 83 | ||
XM_006235504.2 | XP_006235566.1 | 4 - 5 | 609 | 83 | ||
XM_006242839.2 | XP_006242901.1 | 5 - 6 | 690 | 83 | ||
XM_006242840.2 | XP_006242902.1 | 4 - 5 | 609 | 83 | ||
XM_006242841.2 | XP_006242903.1 | 4 - 5 | 609 | 83 | ||
GenBank mRNA | AB121446.1 | - | 561 | 83 |
Gene Symbol: LOC102549510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766105.1 | XP_008764327.1 | 221 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |