Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198755.1 | NP_942050.1 | 1454 | 91 | ||
XM_006235502.2 | XP_006235564.1 | 12 - 13 | 1583 | 91 | ||
XM_006235503.2 | XP_006235565.1 | 11 - 12 | 1502 | 91 | ||
XM_006235504.2 | XP_006235566.1 | 11 - 12 | 1502 | 91 | ||
XM_006242839.2 | XP_006242901.1 | 12 - 13 | 1583 | 91 | ||
XM_006242840.2 | XP_006242902.1 | 11 - 12 | 1502 | 91 | ||
XM_006242841.2 | XP_006242903.1 | 11 - 12 | 1502 | 91 | ||
GenBank mRNA | AB121446.1 | - | 1454 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |