Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128190.1 | NP_001121662.1 | 2 - 3 | 146 | 65 | |
XM_006236007.2 | XP_006236069.1 | 2 - 3 | 179 | 65 | ||
GenBank mRNA | BC158845.1 | - | 2 - 3 | 146 | 7442862 | 65 |
FQ220011.1 | - | 2 - 3 | 158 | 65 | ||
FQ226061.1 | - | 2 - 3 | 156 | 65 | ||
FQ226813.1 | - | 2 - 3 | 156 | 65 | ||
FQ227927.1 | - | 2 - 3 | 141 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |