Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128190.1 | NP_001121662.1 | 6 - 7 | 528 | 131 | |
XM_006236007.2 | XP_006236069.1 | 6 - 7 | 561 | 131 | ||
GenBank mRNA | BC158845.1 | - | 6 - 7 | 528 | 7442862 | 131 |
FQ226061.1 | - | 6 - 7 | 538 | 131 | ||
FQ226813.1 | - | 6 - 7 | 538 | 131 | ||
FQ227927.1 | - | 6 - 7 | 523 | 131 | ||
FQ232170.1 | - | 6 - 7 | 538 | 131 | ||
FQ234730.1 | - | 6 - 7 | 540 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |