Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128190.1 | NP_001121662.1 | 7 - 8 | 587 | 82 | |
XM_006236007.2 | XP_006236069.1 | 7 - 8 | 620 | 82 | ||
GenBank mRNA | BC158845.1 | - | 7 - 8 | 587 | 7442862 | 82 |
FQ226061.1 | - | 7 - 8 | 597 | 82 | ||
FQ226813.1 | - | 7 - 8 | 597 | 82 | ||
FQ227927.1 | - | 7 - 8 | 582 | 82 | ||
FQ232170.1 | - | 7 - 8 | 597 | 82 | ||
FQ234730.1 | - | 7 - 8 | 599 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |