Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014223.2 | NP_001014245.2 | 4 - 5 | 589 | 142 | |
GenBank mRNA | BC079074.1 | - | 4 - 5 | 545 | 7122158 | 142 |
DQ116785.1 | - | 4 - 5 | 589 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |