Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133419.1 | NP_596910.1 | 8 - 9 | 864 | 142 | |
XM_008773596.1 | XP_008771818.1 | 8 - 9 | 1310 | 142 | ||
GenBank mRNA | BC087600.1 | - | 8 - 9 | 823 | 142 | |
BC099832.1 | - | 8 - 9 | 822 | 7934021 | 142 | |
Z34922.1 | - | 8 - 9 | 864 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |