Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173435.1 | NP_001166906.1 | 1 - 2 | 172 | 88 | |
GenBank mRNA | AF176535.1 | - | 1 - 2 | 92 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |