Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106112.1 | NP_001099582.1 | 3 - 4 | 477 | 107 | |
XM_006253927.2 | XP_006253989.1 | 3 - 4 | 262 | 107 | ||
XM_006253928.2 | XP_006253990.1 | 3 - 4 | 200 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |