Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106110.2 | NP_001099580.2 | 4 - 5 | 601 | 96 | |
GenBank mRNA | CV111304.1 | - | 4 - 5 | 309 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |