Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133554.2 | NP_598238.2 | 1410 | 88 | ||
GenBank mRNA | AY102171.1 | - | 1278 | 88 | ||
BC078748.1 | - | 1410 | 7109368 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |