Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271214.1 | NP_001258143.1 | 2 - 3 | 330 | 90 | |
XM_008771657.1 | XP_008769879.1 | 2 - 3 | 335 | 90 | ||
XM_008771658.1 | XP_008769880.1 | 3 - 4 | 649 | 90 | ||
XM_008771659.1 | XP_008769881.1 | 3 - 4 | 719 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |