Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107353.1 | NP_001100823.1 | 7 - 8 | 1147 | 95 | |
XM_006253966.2 | XP_006254028.1 | 7 - 8 | 1224 | 95 | ||
XM_006253967.2 | XP_006254029.1 | 7 - 8 | 971 | 95 | ||
XM_006253968.2 | XP_006254030.1 | 7 - 8 | 1240 | 95 | ||
XM_006253969.2 | XP_006254031.1 | 7 - 8 | 1224 | 95 | ||
XM_008771654.1 | XP_008769876.1 | 7 - 8 | 2408 | 95 | ||
XM_008771655.1 | XP_008769877.1 | 7 - 8 | 2155 | 95 | ||
XM_008771656.1 | XP_008769878.1 | 7 - 8 | 2408 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |