Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020301.1 | NP_064697.1 | 13 - 14 | 1397 | 75 | |
GenBank mRNA | X66140.1 | - | 13 - 14 | 1397 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |