Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020301.1 | NP_064697.1 | 1 - 2 | 96 | 93 | |
GenBank mRNA | X66140.1 | - | 1 - 2 | 96 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |