Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108770.1 | NP_001102240.1 | 1 - 2 | 99 | 77 | |
XM_006243332.2 | XP_006243394.1 | 1 - 2 | 83 | 77 | ||
XM_006243337.2 | XP_006243399.1 | 1 - 2 | 122 | 77 | ||
XM_006243341.2 | XP_006243403.1 | 1 - 2 | 122 | 77 | ||
XM_008766317.1 | XP_008764539.1 | 1 - 2 | 83 | 77 | ||
XM_008766318.1 | XP_008764540.1 | 1 - 2 | 83 | 77 | ||
XM_008766319.1 | XP_008764541.1 | 1 - 2 | 122 | 77 | ||
XM_008766320.1 | XP_008764542.1 | 1 - 2 | 83 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |