Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053896.2 | NP_446348.2 | 2 - 3 | 228 | 62 | |
GenBank mRNA | BC098910.1 | - | 2 - 3 | 220 | 7455646 | 62 |
BE111477.1 | - | 2 - 3 | 228 | 62 | ||
U60063.1 | - | 2 - 3 | 224 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |