Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758841.1 | XP_008757063.1 | 4 - 5 | 729 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |