Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250518.2 | XP_006250580.1 | 2 - 3 | 561 | 142 | |
XM_006250519.2 | XP_006250581.1 | 561 | 142 | |||
XM_006250520.2 | XP_006250582.1 | 2 - 3 | 561 | 142 | ||
XM_006250522.2 | XP_006250584.1 | 2 - 3 | 561 | 142 | ||
XM_008769957.1 | XP_008768179.1 | 23 - 24 | 3727 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |