Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013987.1 | NP_001014009.1 | 7 - 8 | 527 | 83 | |
XM_006250547.2 | XP_006250609.1 | 7 - 8 | 652 | 83 | ||
XM_006250548.2 | XP_006250610.2 | 7 - 8 | 642 | 83 | ||
XM_008769928.1 | XP_008768150.1 | 7 - 8 | 642 | 83 | ||
XM_008769929.1 | XP_008768151.1 | 7 - 8 | 642 | 83 | ||
XM_008769930.1 | XP_008768152.1 | 5 - 6 | 313 | 83 | ||
GenBank mRNA | BC088426.1 | - | 7 - 8 | 527 | 7123983 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |