Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191101.1 | NP_001178030.1 | 5 - 6 | 806 | 144 | |
XM_006250582.2 | XP_006250644.1 | 6 - 7 | 882 | 144 | ||
XM_008769943.1 | XP_008768165.1 | 6 - 7 | 1165 | 144 | ||
XM_008769944.1 | XP_008768166.1 | 7 - 8 | 984 | 144 | ||
XM_008769945.1 | XP_008768167.1 | 5 - 6 | 1623 | 144 | ||
XM_008769946.1 | XP_008768168.1 | 5 - 6 | 1623 | 144 | ||
XM_008769947.1 | XP_008768169.1 | 5 - 6 | 1623 | 144 | ||
XM_008769948.1 | XP_008768170.1 | 5 - 6 | 1623 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |