Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105994.1 | NP_001099464.1 | 5 - 6 | 671 | 150 | |
XM_006250541.1 | XP_006250603.1 | 5 - 6 | 724 | 150 | ||
XM_008769927.1 | XP_008768149.1 | 5 - 6 | 1583 | 150 | ||
GenBank mRNA | FQ213206.1 | - | 5 - 6 | 597 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |