Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022668.2 | NP_073159.2 | 5 - 6 | 672 | 83 | |
XM_006255867.2 | XP_006255929.1 | 5 - 6 | 1145 | 83 | ||
XM_008772687.1 | XP_008770909.1 | 5 - 6 | 1149 | 83 | ||
GenBank mRNA | BC087717.1 | - | 5 - 6 | 672 | 7317044 | 83 |
FQ214199.1 | - | 5 - 6 | 652 | 83 | ||
L21995.1 | - | 4 - 5 | 503 | 83 | ||
M99485.1 | - | 5 - 6 | 672 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |