Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009176.2 | NP_001009176.1 | 4 - 5 | 1137 | 140 | |
GenBank mRNA | BC129078.1 | - | 4 - 5 | 1137 | 8371876 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |