Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191731.2 | NP_001178660.2 | 2 - 3 | 331 | 148 | |
XM_006257271.2 | XP_006257333.1 | 3 - 4 | 416 | 148 | ||
XM_008773409.1 | XP_008771631.1 | 2 - 3 | 346 | 148 | ||
XM_008773410.1 | XP_008771632.1 | 3 - 4 | 472 | 148 | ||
XM_008773411.1 | XP_008771633.1 | 3 - 4 | 469 | 148 | ||
XM_008773412.1 | XP_008771634.1 | 3 - 4 | 349 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |