Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024906.1 | NP_001020077.1 | 1 - 2 | 111 | 105 | |
GenBank mRNA | BC097448.1 | - | 1 - 2 | 111 | 7456585 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |