Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107412.1 | NP_001100882.1 | 4 - 5 | 398 | 110 | |
XM_006255141.2 | XP_006255203.1 | 3 - 4 | 4816 | 110 | ||
XM_006255143.1 | XP_006255205.1 | 4 - 5 | 400 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |