Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025721.1 | NP_001020892.1 | 4 - 5 | 371 | 71 | |
GenBank mRNA | BC097495.1 | - | 4 - 5 | 371 | 7461783 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |