Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022701.2 | NP_073192.2 | 9 - 10 | 1079 | 120 | |
XM_006256024.2 | XP_006256086.1 | 9 - 10 | 1098 | 120 | ||
GenBank mRNA | BC064652.1 | - | 9 - 10 | 1078 | 5598180 | 120 |
CB324456.1 | - | 4 - 5 | 120 | |||
CB326388.1 | - | 4 - 5 | 120 | |||
U60976.1 | - | 9 - 10 | 1009 | 120 | ||
U60977.1 | - | 8 - 9 | 1083 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |