Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212498.1 | NP_997663.1 | 11 - 12 | 1062 | 104 | |
XM_006255948.2 | XP_006256010.1 | 10 - 11 | 1028 | 104 | ||
XM_008772731.1 | XP_008770953.1 | 9 - 10 | 956 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |