Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166275.1 | NP_001159747.1 | 15 - 16 | 3746 | 99 | |
GenBank mRNA | BC085955.1 | - | 15 - 16 | 3746 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |